Anti-IgG Goat Polyclonal Antibody [clone: IgG] (trFluor™ Eu)
Leverancier: AAT Bioquest
Certificaten
Over dit artikel
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
- Large stokes shift
- Improved stability
- High emission yield
- Long emission half-life
Caution: For research use only.
Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: trFluor™ Eu
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Specificaties
- Catalog No:
- AATB16847
- AATB16725
- Antigen symbol:
- IgG
- IgG
- Antigen name:
- Immunoglobulin G
- Immunoglobulin G
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- trFluor™ Eu
- trFluor™ Eu
- Clone:
- IgG
- IgG
- Reactivity:
- Rabbit
- Rabbit
- Host:
- Goat
- Goat
- Isotype:
- IgG (H, L)
- IgG (H, L)
- Cross adsorption:
- Min×Human
- Min×Human
Specificaties
Frequently Bought Together