Order Entry
Netherlands
ContactUsLinkComponent
FMOC-Asp(5/6-TAMRA)-OH for biomolecular labeling
Irritant
FMOC-Asp(5/6-TAMRA)-OH for biomolecular labeling
Catalog # AATB5005
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
FMOC-Asp(5/6-TAMRA)-OH for biomolecular labeling
Catalog # AATB5005
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
ADR:5,II
 :GHS07