Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Carboxyrhodamine 110 (5-CR110)
5-Carboxyrhodamine 110 (5-CR110) single isomer ≥95%
5-Carboxyrhodamine 110 (5-CR110) single isomer ≥95%
5-Carboxyrhodamine 110 (5-CR110) single isomer ≥95%
Catalog # AATB322
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
5-Carboxyrhodamine 110 (5-CR110) single isomer ≥95%
Catalog # AATB322
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
410.81 g/mol
Product Details