
Tide Fluor™ 8WS, succinimidyl ester, Tide Fluor™ fluorescent probe
Catalog # AATB2338
Leverancier: AAT Bioquest
CAS-nummer:
Specificaties
- Pack type:Plastic bottle
- Pk:1 mg
- Seq:2070
Specificaties
Over dit artikel
:
:1343.73 g/mol
ADR:5,II
:GHS07