
N,N,N-Trimethyl-4-(6-phenyl-1,3,5-hexatrien-1-yl)phenylammonium-p-toluenesulphonate ≥95%
Catalog # AATB21492
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:461.62 g/mol
ADR:5,II
:GHS07
CAS Number:115534-33-3
Formula:C₂₈H₃₁NO₃S