Rhod-FF AM
Supplier: AAT Bioquest
Jump to:
About this item
Calcium measurement is critical for numerous biological investigations.
- Calcium-dependent fluorescence
- Low binding affinity
- Cell permeant
: For research use only.
- Molecular Weight:1145.9 g/mol
- ADR:5,II
2 Options Available Below
Product Details & Documents
Calcium measurement is critical for numerous biological investigations.
- Calcium-dependent fluorescence
- Low binding affinity
- Cell permeant
: For research use only.
- Molecular Weight:1145.9 g/mol
- ADR:5,II



