
Calbryte™ 520 AM
Supplier: AAT BIOQUEST
Certificates
About this item
:
:1090.9 g/mol
ADR:5,II
:GHS07
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Accessories
Supplier: Merck
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® PureStep, Multi-Layer Adhesive Floor Mats
Supplier: VWR Collection
VWR®, Cryovials
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, PharMed® BPT, Avantor®
Supplier: Avantor Fluid Handling
Hypodermic safety needles, Agani™
Supplier: TERUMO
Fluo-8 AM
Supplier: AAT BIOQUEST
HUMAN PRO-COLLAGEN I ALPHA 1 ELISA KIT 1 * 96 Tests
Supplier: Abcam