Order Entry
Netherlands
ContactUsLinkComponent
Cal Red™ R525/650 potassium salt
Irritant
Cal Red™ R525/650 potassium salt
Catalogus # AATB20588
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Cal Red™ R525/650 potassium salt
Catalogus # AATB20588
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 µG
  • Seq:
    1370

Specifications

About this item

 :
ADR:5,II
 :1000 g/mol
 :GHS07