Order Entry
Netherlands
ContactUsLinkComponent
N-Succinimidyl 6-(2,4-Dinitroanilino)hexanoate ≥90%
Irritant
N-Succinimidyl 6-(2,4-Dinitroanilino)hexanoate ≥90%
Catalog # AATB2021
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
N-Succinimidyl 6-(2,4-Dinitroanilino)hexanoate ≥90%
Catalog # AATB2021
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :394.34 g/mol
ADR:5,II
 :GHS07
CAS Number:82321-04-8
Formula:C₁₆H₁₈N₄O₈
MDL Number:MFCD00191630