Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Red 780 succinimidyl ester
Red 780 succinimidyl ester, mFluor™ fluorescent probe
Red 780 succinimidyl ester, mFluor™ fluorescent probe
Irritant
Red 780 succinimidyl ester, mFluor™ fluorescent probe
Catalog # AATB1191
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Irritant
Red 780 succinimidyl ester, mFluor™ fluorescent probe
Catalog # AATB1191
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
1328.65 g/mol
ADR:
5,II
:
GHS07
Product Details