Recombinant Human PDIA6 (from E. coli), His-tag
ABNOP7940.100UG
: Abnova
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:Q15084
- Protein/peptide name:PDIA6
- Sequence:20a.a.-440a.a.