Recombinant Human UBE2G1 (from E. coli), His-tag
Catalogus # ABNOP7740.500UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:500 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:NP_003333
- Protein/peptide name:UBE2G1
- Sequence:1a.a.-170a.a