Order Entry
Netherlands
ContactUsLinkComponent
Recombinant Human PDGFC (from HEK293 cells)
Recombinant Human PDGFC (from HEK293 cells)
Catalog # ABNOP7348.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Human PDGFC (from HEK293 cells)
Catalog # ABNOP7348.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Gene ID:
    Q9NRA1
  • Protein/peptide name:
    PDGFC
  • Sequence:
    235a.a.-345a.a

Specifications

About this item