Anti-PCNA1 (Proliferating cellular nuclear antigen 1) conjugated to KLH Rabbit Recombinant Antibody [clone: SAIC-27C-101]
Catalog # ABNORAB03735.200UG
Supplier: Abnova
Specifications
- Antigen symbol:PCNA1 (Proliferating cellular nuclear antigen 1) conjugated to KLH
- Clonality:Recombinant
- Clone:SAIC-27C-101
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide corresponding to human PCNA1 (Proliferating cellular nuclear antigen 1) conjugated to KLH.
- Pk:200 µG