Anti-Notch 2 Mouse Recombinant Antibody [clone: B9]
Catalog # ABNORAB03726.100UG
Supplier: Abnova
Specifications
- Antigen symbol:Notch 2
- Clonality:Recombinant
- Clone:B9
- Host:Mouse
- Reactivity:Mouse
- Immunogen:Recombinant mouse monoclonal antibody raised against NRR of mouse Notch 2.
- Pk:100 µG