Anti-MDM2 (pSer166) Rabbit Recombinant Antibody [clone: R08-8C7] ()
Catalog # ABNORAB01668.100UL
Supplier: Abnova
Specifications
- Antigen symbol:MDM2
- Clonality:Recombinant
- Clone:R08-8C7
- Host:Rabbit
- Reactivity:Human
- Modification:pSer166
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic phosphopeptide corresponding to residues surrounding Ser166 of human MDM2
- Pk:100 µl