Anti-mtTFA Rabbit Recombinant Antibody [clone: R08-3E2]
ABNORAB01728.100UL
: Abnova
- Antigen symbol:mtTFA
- Clonality:Recombinant
- Clone:R08-3E2
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human mtTFA
- Pk:100 µl