Anti-Midkine Rabbit Recombinant Antibody [clone: R06-4B2]
Catalog # ABNORAB01743.100UL
Supplier: Abnova
Specifications
- Antigen symbol:Midkine
- Clonality:Recombinant
- Clone:R06-4B2
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Midkine
- Pk:100 µl