Anti-ZNF364 Rabbit Recombinant Antibody [clone: R06-1B3]
Catalog # ABNORAB01744.100UL
Supplier: Abnova
Specifications
- Antigen symbol:ZNF364
- Clonality:Recombinant
- Clone:R06-1B3
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human ZNF364
- Pk:100 µl