Anti-GSK3 alpha Rabbit Recombinant Antibody [clone: R08-8G7]
Catalog # ABNORAB01676.100UL
Supplier: Abnova
Specifications
- Antigen symbol:GSK3 alpha
- Clonality:Recombinant
- Clone:R08-8G7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human GSK3 alpha
- Pk:100 µl