Anti-CHEK1 (pSer317) Rabbit Monoclonal Antibody [clone: G1] (APC)
Catalogus # ABNOMAB23477.100
Supplier: Abnova
Specifications
- Antigen symbol:CHEK1
- Clonality:Monoclonal
- Clone:G1
- Conjugation:APC (Allophycocyanin)
- Host:Rabbit
- Reactivity:Human
- Modification:pSer317
- Immunogen:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human CHEK1
- Pk:100 AU