Anti-CRKL (pTyr207) Rabbit Monoclonal Antibody [clone: G4] (APC)
Catalog # ABNOMAB23433.100
Supplier: Abnova
Specifications
- Antigen symbol:CRKL
- Clonality:Monoclonal
- Clone:G4
- Conjugation:APC (Allophycocyanin)
- Host:Rabbit
- Reactivity:Human
- Modification:pTyr207
- Immunogen:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human CRKL
- Pk:100 AU