Safety Shoes, Clogs, Active, 87020
: Abeba
Anti-static safety clogs made from grained leather with a non foldable, adjustable heel strap and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile.
- Increased breathability
- Increased durability of the upper
- Easy-clean upper
- Sole resistance to acids and bases
- Adjustable instep strap with buckle fastening
- Leather - tear resistance in requirements not less than 120 N
- Lining - increased water vapor permeability in the requirements of mininum 2,0 mg
Suitable for use in gastronomy/kitchen, horeca, hospitals, pharmaceuticals.
Colour: White, Toe: Thermo plastic resistant to deformation, light, elastic.
Slip resistance on ceramic, coated with SLS solution and on steel, coated with glycerol - slip resistance on ceramic substrate, covered with SLS solution and on steel substrate, covered with glycerol. Antistatic footwear - antistatic properties. Energy absorption in the heel area - energy absorbtion in the heel area. The sole resistant to diesel fuel - sole resistance to diesel fuel.
: EN ISO 20347:2012; 19/2022/PPE/1439/B edition 1; OB (Basic requirements met); SRC (Slip resistance on ceramic).
Frequently Bought Together