Anti-NPFF Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-8150R-A750
: Bioss
- Antibody type:Primary
- Antigen name:NPFF
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Epitope:51-113/113
- Form:Liquid
- Gene ID:8620
- Antigen synonyms:Neuropeptide FF amide peptide precursor|NPFF protein|NPFF_HUMAN|NPFF|FMRFamide related peptides precursor|FMRFAL|FMRFamide-related peptides [Precursor]|FMFRamide related peptide precursor
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:5,3 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human NPFF/FMRFamide
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
FMRFamide related peptides are morphine modulating peptides that have many physiologic effects, including the modulation of morphine induced analgesia, elevation of arterial blood pressure, and increased somatostatin secretion from the pancreas. Neuropeptide FF potentiates and sensitises ACCN2 and ACCN3 channels.
Type: Primary
Antigen: NPFF
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-113/113
Host: Rabbit
Isotype: IgG
Reactivity: