Anti-beta 2 Microglobulin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6294R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:beta 2 Microglobulin
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Epitope:51-119/119
- Form:Liquid
- Gene ID:567
- Antigen synonyms:Beta-2-MG|Hdcma22p|Beta chain of mhc class 1 proteins|Beta 2 microglobulin precursor|B2M|beta 2 Microglobulin|B2MG_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:12 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human beta 2 Microglobulin corresponding to amino acids 51-97.
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This gene encodes a serum protein found in association with the major histocompatibility complex (MHC) class I heavy chain on the surface of nearly all nucleated cells. The protein has a predominantly beta-pleated sheet structure that can form amyloid fibrils in some pathological conditions. A mutation in this gene has been shown to result in hypercatabolic hypoproteinemia.
Type: Primary
Antigen: beta 2 Microglobulin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-119/119
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse