Bestelling invoeren
Netherlands
ContactUsLinkComponent
 
Anti-MMP-26 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-1377R-CY3
 :  Bioss
Anti-MMP-26 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-1377R-CY3
 :  Bioss

 

  • Type antilichaam:
    Primair
  • Clonality:
    Polyklonaal
  • Conjugatie:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemie:
    Yes
  • ImmunoFluorescentie:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitoop:
    185-261/261
  • Formulier:
    Liquid
  • Gen-ID:
    56547
  • Antigeen synoniemen:
    Matrilysin 2|Endometase|MMP26|MMP26_HUMAN| Matrix Metalloproteinase 26|MMP 26.
  • Modification:
    Unmodified
  • Opslagbuffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Moleculair gewicht:
    18 kDa
  • Opslagtemperatuur:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentratie:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogeen:
    KLH conjugated synthetic peptide derived from human MMP26 intermedial
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Verpakking:
    Vial
  • VPE:
    100 µl

 

 

Matrix metalloproteinase 26 preprotein; gelatinase A; 70kD type IV collagenase; gelatinase neutrophil. Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes as well as in disease processes. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. MMP26 degrades type IV collagen, the major structural component of basement membranes. The enzyme plays a role in endometrial menstrual breakdown, regulation of vascularization and the inflammatory response.Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodelling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. MMP26, also known as Matrilysin 2, was first cloned from human fetal cells, and identified as an MMP most closely related to MMP7 (Matrilysin 1). The homology between MMP7 and MMP26 is low (only 38% identical), thus the functions are unlikely to be similar. Homology is much higher (48% identical) for the comparable region of MMP12, but MMP26 appears to have broader substrate specificity than does MMP12. MMP26, like MMP7, lacks the hemopexin domain common to the other MMPs, but contains a Propeptide domain, cysteine switch activation site, followed by a catalytic domain, and a short vestige of the hinge region. MMP26 is apparently not glycosylated, and is a secreted MMP. Tissue analysis shows MMP26 most strongly in placenta and uterus, but also in kidney cells, lung cells, lymphocytes and lung or endometrial carcinoma cells. MMP26 is proteolytically active, cleaving casein in zymograms, and gelatin, a1PI, fibrinogen, fibronectin, vitronectin, type IV collagen, and apparently activating MMP9.

Type: Primary
Antigen: MMP-26
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 185-261/261
Host: Rabbit
Isotype: IgG
Reactivity: Human