Anti-AIF1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-1363R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:AIF1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:51-147/147
- Form:Liquid
- Gene ID:199
- Antigen synonyms:AIF-1|IRT-1|Protein G1|IRT1|Ionized calcium-binding adapter molecule 1|Allograft inflammatory factor 1|AIF1|IBA1|G1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:16 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human AIF1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Actin-binding protein that enhances membrane ruffling and RAC activation. Enhances the actin-bundling activity of LCP1. Binds calcium. Plays a role in RAC signaling and in phagocytosis. May play a role in macrophage activation and function. Promotes the proliferation of vascular smooth muscle cells and of T-lymphocytes. Enhances lymphocyte migration. Plays a role in vascular inflammation.
Type: Primary
Antigen: AIF1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:51-147/147
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat