Anti-CRHR2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-2792R-A750
: Bioss
- Antibody type:Primary
- Antigen name:CRHR2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:51-150/411
- Form:Liquid
- Gene ID:1395
- Antigen synonyms:CRF-R-2|Corticotropin-releasing factor receptor 2|CRH2R|CRF2|CRHR2|CRF2R|CRH-R2|HM-CRF|CRFR2|Corticotropin-releasing hormone receptor 2|CRFR-2|CRF-R2|CRF-RB|CRH-R-2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:43 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CRHR2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
G-protein coupled receptor for CRH (corticotropin-releasing factor), UCN (urocortin), UCN2 and UCN3. Has high affinity for UCN. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G proteins) and down-stream effectors, such as adenylate cyclase. Promotes the activation of adenylate cyclase, leading to increased intracellular cAMP levels.
Type: Primary
Antigen: CRHR2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/411
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse