Anti-Semaphorin 7A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2702R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Semaphorin 7A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:351-450/666
- Form:Liquid
- Gene ID:8482
- Antigen synonyms:CDw108|CD108|Semaphorin-K1|H-SEMA-K1|JMH|Semaphorin-7A|Sema K1|Sema L|H-Sema-L|Semaphorin-L|SEMAL|John-Milton-Hargen human blood group Ag|SEMAK1|SEMA7A|JMH blood group antigen
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:66 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Semaphorin 7A
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Plays an important role in integrin-mediated signaling and functions both in regulating cell migration and immune responses. Promotes formation of focal adhesion complexes, activation of the protein kinase PTK2/FAK1 and subsequent phosphorylation of MAPK1 and MAPK3. Promotes production of proinflammatory cytokines by monocytes and macrophages. Plays an important role in modulating inflammation and T-cell-mediated immune responses. Promotes axon growth in the embryonic olfactory bulb. Promotes attachment, spreading and dendrite outgrowth in melanocytes.
Type: Primary
Antigen: Semaphorin 7A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 351-450/666
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat