Human Type AB Serum (HCV, HIV, HBsAG Free), MP Biomedicals
Catalog # ICNA092931949
Supplier: MP Biomedicals
Specifications
- Omschrijving:Human Type AB serum (HCV, HIV, HBsAG free)
- VPE:100 mL
Specifications
About this item
Human Type AB Serum is used as a growth supplement for cell culture media and in other research applications.
- Collected from Type AB human blood
- Liquid media
- Store at —20 °C
It has been proven to grow many human cell lines at a faster rate and with a smaller percentage of serum than mixed blood group serum.