6-Bromo-3-fluoro-2-(trifluoromethyl)aniline ≥95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1805152-52-6
:
Formula:C₇H₄BrF₄N
MDL Number:MFCD28740806
:258.01097 g/mol
UN:2810
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Ethylene glycol, high purity
Supplier: VWR Chemicals
Acetonitrile ≥99.7%, Environmental Grade
Supplier: Thermo Fisher Scientific
N-Bromosuccinimide 99%
Supplier: Thermo Fisher Scientific
Methanol 99.8%, ECD tested for pesticide analysis for residue analysis
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals