Order Entry
Netherlands
Orders LinkContactUsLinkComponent
2-Bromo-3-hydroxypyridine ≥95%

2-Bromo-3-hydroxypyridine ≥95%

Supplier:   Apollo Scientific
2-Bromo-3-hydroxypyridine ≥95%
Supplier:  Apollo Scientific
Certificates
2-Bromo-3-hydroxypyridine ≥95%

2 Options Available Below

Product Details & Documents
Synonyms:  2-Bromo-3-pyridinol
  • CAS Number:6602-32-0
  • Formula:C₅H₄BrNO
  • MDL Number:MFCD00006220
  • Molecular Weight:174 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 21,80 €
    21,80 €
    1 * 25 g
       21,80 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 65,20 €
    65,20 €
    1 * 100 g
       65,20 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.