Methyl ethyl ketone, liquid, Burdick & Jackson™
Catalog # HONC65806-200L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Steel drum
- Pk:200 L
- Seq:5549
Specifications
About this item
:
Boiling Pt:79 - 81 °C
Boiling Pt:79…81 °C
Melting Pt:-87 °C
Melting Pt:–87 °C
Density:0.805 g/cm3 (20 °C)
Flash Pt:-4 °C
UN:1193
ADR:3,II
:72.11 g/mol
CAS Number:78-93-3
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648
Flash Pt:–4 °C