Order Entry
Netherlands
ContactUsLinkComponent
Indigo carmine solution for PANPEHA indicator paper, Riedel-de Haën™
Indigo carmine solution for PANPEHA indicator paper, Riedel-de Haën™
Catalog # HONC34661-2.5L
CAS Number:  
Indigo carmine solution for PANPEHA indicator paper, Riedel-de Haën™
Catalog # HONC34661-2.5L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :466.36 g/mol
CAS Number:860-22-0
Formula:C₁₆H₈N₂Na₂O₈S₂
MDL Number:MFCD00005723