Chlorophenol red solution for PANPEHA indicator paper, Riedel-de Haën™
Catalog # HONC34658-5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic can
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:423.27 g/mol
CAS Number:4430-20-0
Formula:C₁₉H₁₂Cl₂O₅S
MDL Number:MFCD00005877