Order Entry
Netherlands
ContactUsLinkComponent
SDAD (NHS-SS-Diazirine) (succinimidyl 2-((4,4'-azipentanamido)ethyl)-1,3'-dithiopropionate), Pierce™
SDAD (NHS-SS-Diazirine) (succinimidyl 2-((4,4'-azipentanamido)ethyl)-1,3'-dithiopropionate), Pierce™
Catalog # PIER26169
CAS Number:  
undefined
SDAD (NHS-SS-Diazirine) (succinimidyl 2-((4,4'-azipentanamido)ethyl)-1,3'-dithiopropionate), Pierce™
Catalog # PIER26169
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    NHS Ester, Diazirine
  • Spacer:
    Cleavable, Long
  • Form:
    Solid
  • Quantity:
    50 mg
  • Type:
    Crosslinker
  • Solubility:
    DMF, DMSO
  • Spacer Arm:
    13.5 Å

Specification Test Results

About this item

 :
 :388.46 g/mol
Storage Temperature:Refrigerator
CAS Number:1253202-38-8
Formula:C₁₄H₂₀N₄O₅S₂