
BMB (1,4-Bis(maleimido)butane), Pierce™
PIER22331
:
- Pk:50 mg
- Seq:2370
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Homobifunctional
- Reactive Moiety:Maleimide
- Spacer:Mid-Length
- Form:Solid
- Quantity:50 mg
- Type:Crosslinker
- Solubility:DMF, DMSO
- Spacer Arm:10.9 Å
:
:248.24 g/mol
:GHS07
:Refrigerator
:28537-70-4
:C₁₂H₁₂N₂O₄
:MFCD01318342