Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Ticagrelor, Reference Standards

Supplier:   USP
Ticagrelor, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1667133.
  • 1667177.
  • 1667188.
  • Description:
  • Ticagrelor Related Compound B
  • Ticagrelor
  • Ticagrelor Related Compound A
  • Synonym(s):
  • (1R,2S)-2-(3,4-difluorophenyl)cyclopropan-1-amine (R)-mandelate
  • CAS No.:
  • 1788033-05-5
  • 274693-27-5
  • 376608-71-8
  • UN No.:
  • 3077
  • 2811

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1667133.
    Ticagrelor Related Compound B
    Ticagrelor Related Compound B
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 940,00 €
    1 940,00 €
    1 * 25 mg
       1 940,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1667133.
    Description
    Ticagrelor Related Compound B
    Synonym(s)
    CAS No.
    1788033-05-5
    UN No.
  • 1667177.
    Ticagrelor
    Ticagrelor
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 2 140,00 €
    2 140,00 €
    1 * 200 mg
       2 140,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1667177.
    Description
    Ticagrelor
    Synonym(s)
    CAS No.
    274693-27-5
    UN No.
    3077
  • 1667188.
    Ticagrelor Related Compound A
    Ticagrelor Related Compound A
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 640,00 €
    1 640,00 €
    1 * 25 mg
       1 640,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1667188.
    Description
    Ticagrelor Related Compound A
    Synonym(s)
    (1R,2S)-2-(3,4-difluorophenyl)cyclopropan-1-amine (R)-mandelate
    CAS No.
    376608-71-8
    UN No.
    2811