Order Entry
Netherlands
ContactUsLinkComponent
Sulpho-EGS (Ethylene glycol bis(sulphosuccinimidyl succinate)), Pierce™
Sulpho-EGS (Ethylene glycol bis(sulphosuccinimidyl succinate)), Pierce™
Catalog # PIER21566
CAS Number:  
undefined
Sulpho-EGS (Ethylene glycol bis(sulphosuccinimidyl succinate)), Pierce™
Catalog # PIER21566
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Homobifunctional
  • Reactive Moiety:
    Sulfo-NHS Ester
  • Spacer:
    Cleavable, Long
  • Form:
    Powder
  • Quantity:
    50 mg
  • Type:
    Crosslinker
  • Solubility:
    Water
  • Spacer Arm:
    16.1 Å

Specification Test Results

About this item

 :
 :660.45 g/mol
Storage Temperature:Freezer
CAS Number:67410-92-6
Formula:C₁₈H₁₈O₁₈N₂Na₂S₂
MDL Number:MFCD00054988