SIAB (N-Succinimidyl (4-iodoacetyl)aminobenzoate)
Catalog # BC19
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Spacer Arm (Å):10.6
- Reactive Toward:Primary Amine + Sulfhydryl
- Membrane Permeable:Yes
- Water Soluble:No
- Cleavable/ Reversible:No
Specification Test Results
About this item
:
:400.125 g/mol
CAS Number:72252-96-1
Formula:C₁₃H₉IN₂O₅
MDL Number:MFCD00078537