Order Entry
Netherlands
ContactUsLinkComponent
MBS (N-Succinimidyl 3-maleimidobenzoate)
Irritant
MBS (N-Succinimidyl 3-maleimidobenzoate)
Catalog # BC11
Supplier:  G-Biosciences
CAS Number:  
undefined
Irritant
MBS (N-Succinimidyl 3-maleimidobenzoate)
Catalog # BC11
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Spacer Arm (Å):
    9.9
  • Reactive Toward:
    Primary Amine + Sulfhydryl
  • Membrane Permeable:
    Yes
  • Water Soluble:
    No
  • Cleavable/ Reversible:
    No

Specification Test Results

About this item

 :
 :314.25 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:58626-38-3
Formula:C₁₅H₁₀N₂O₆
MDL Number:MFCD00005514