Order Entry
Netherlands
ContactUsLinkComponent
Rotor package, MLN-80 NVT, Optima
Rotor package, MLN-80 NVT, Optima
  521-4042
undefined
Rotor package, MLN-80 NVT, Optima
  521-4042
  • Description:
    MLN-80 NVT rotor package, 80000 rpm, 389000 ×g, 8×8 ml. Major applications: Rapid contamination-free isopycnic isolation of plasmid DNA. For use in Optima MAX-XP benchtop ultracentrifuges and Optima MAX ultracentrifuges
  • Pk:
    1
  • For:
    Near vertical (ultracentrifugation)
  • Max. RPM:
    80000
  • No. of tubes accepted:
    8×8 ml
  • Radius:
    54,2 mm