Order Entry
Netherlands
ContactUsLinkComponent
Sulpho-SANPAH (Sulphosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate), No-Weigh™ Format, Pierce™
Irritant
Sulpho-SANPAH (Sulphosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate), No-Weigh™ Format, Pierce™
Catalog # PIERA35395
CAS Number:  
Irritant
Sulpho-SANPAH (Sulphosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate), No-Weigh™ Format, Pierce™
Catalog # PIERA35395
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275
  • Pk Info:
    10 x 1 mg

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    Sulfo-NHS Ester, Aryl Azide
  • Spacer:
    Long
  • Form:
    Solid
  • Quantity:
    50 mg
  • Type:
    Crosslinker
  • Solubility:
    Water

Specification Test Results

About this item

 :
 :492.40 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:102568-43-4
Formula:C₁₆H₁₇O₉N₆NaS