Order Entry
Netherlands
ContactUsLinkComponent
SMPH (Succinimidyl-6-((beta-maleimidopropionamido)hexanoate), Pierce™
SMPH (Succinimidyl-6-((beta-maleimidopropionamido)hexanoate), Pierce™
Catalog # PIER22363
CAS Number:  
undefined
SMPH (Succinimidyl-6-((beta-maleimidopropionamido)hexanoate), Pierce™
Catalog # PIER22363
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    Maleimide, NHS Ester
  • Spacer:
    Long
  • Form:
    Powder
  • Quantity:
    50 mg
  • Type:
    Crosslinker
  • Solubility:
    DMF, DMSO

Specification Test Results

About this item

 :
 :379.36 g/mol
Storage Temperature:Refrigerator
CAS Number:367927-39-7
Formula:C₁₇H₂₁N₃O₇
MDL Number:MFCD01863460