Order Entry
Netherlands
ContactUsLinkComponent
MPBH (4-(4-N-Maleimidophenyl)butyric acid hydrazide hydrochloride), Pierce™
MPBH (4-(4-N-Maleimidophenyl)butyric acid hydrazide hydrochloride), Pierce™
Catalog # PIER22305
CAS Number:  
undefined
MPBH (4-(4-N-Maleimidophenyl)butyric acid hydrazide hydrochloride), Pierce™
Catalog # PIER22305
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    Amine, Hydrazide, Maleimide
  • Spacer:
    Long
  • Form:
    Powder
  • Quantity:
    50 mg
  • Type:
    Crosslinker
  • Solubility:
    DMF, DMSO

Specification Test Results

About this item

 :
 :309.75 g/mol
Storage Temperature:Refrigerator
Formula:C₁₄H₁₆N₃ClO₃