
Posaconazole
Catalog # ACRO467660010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to light yellow to beige powder or crystals
- Infrared spectrum:Conforms
- Melting point:165°C to 170°C
- Water:≤ 1%
- Specific optical rotation:-24° to -30° (20°C, 589nm) (1% in CHCl3)
Specification Test Results
About this item
:
:700.79 g/mol
Melting Pt:170°C…179°C
:GHS08
Storage Temperature:Refrigerator
CAS Number:171228-49-2
Formula:C₃₇H₄₂F₂N₈O₄
MDL Number:MFCD00941162