

β-Methasone sodium phosphate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:151-73-5
:
Formula:C₂₂H₂₈FO₈P·₂Na
MDL Number:MFCD00200361
:516.41 g/mol
Melting Pt:206°C…211°C
EINECS:205-797-0
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to almost white powder
- Infrared spectrum:Conforms
- Water:≤ 10%
- HPLC:≥ 96.0%
- Specific optical rotation:+98° to +105° (20°C, 589nm) (1% in water)
Specification Test Results
Frequently Bought Together