Order Entry
Netherlands
ContactUsLinkComponent
Ammonium nitrate 50 g/L (99,999%) in water, ARISTAR® matrix modifier solution, for graphite furnace AAS
Ammonium nitrate 50 g/L (99,999%) in water, ARISTAR® matrix modifier solution, for graphite furnace AAS
Catalog # 91277.154
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium nitrate 50 g/L (99,999%) in water, ARISTAR® matrix modifier solution, for graphite furnace AAS
Catalog # 91277.154
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :80.04 g/mol
Storage Temperature:Ambient
CAS Number:6484-52-2
Formula:NH₄NO₃
MDL Number:MFCD00011425