Order Entry
Netherlands
ContactUsLinkComponent
Anti-POU2F3 Rabbit Polyclonal Antibody
Anti-POU2F3 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-83966
Supplier:  Novus Biologicals
Anti-POU2F3 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-83966
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    POU2F3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    25833
  • Antigen synonyms:
    POU transcription factor|MGC126698|Skn-1a|class 2|oct-11|Epoc-1|POU class 2 homeobox 3|OCT11FLJ40063|Octamer-binding transcription factor 11|Transcription factor Skn-1|PLA-1|OTF11|transcription factor 3|POU domain class 2|POU domain|Octamer-binding protein 11|PLA1|Transcription factor PLA-1|OTF-11
  • Storage buffer:
    PBS (pH 7,2) and 40% Glycerol
  • Storage temperature:
    Store at 4 °C short term. Aliquot and store at -20 °C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    This antibody was developed against Recombinant Protein corresponding to amino acids:LESMHTDIKMSGDVADSTDARSTLSQVEPGNDRNGLDFNRQIKTEDLSDSLQQTLSHRPCHLSQGPAMMSGNQMSGLNASPCQDMASLHPLQQLVLVPGHLQSVSQFLLSQTQP
  • Purification:
    Immunogen affinity purified
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

The POU2F3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to POU2F3. This antibody reacts with human. The POU2F3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: POU2F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human