Corning® Lymphocyte serum-free medium
Supplier: Corning
Certificates
About this item
The serum-free medium is used to support the growth of T-cells activated with immobilized anti-CD3 antibody. Interleukin-2 is a necessary component for a successful culture. This formulation has been shown to work with the following cell types: CIK/DC-CIK cell lines, T-cells, CAR-T, TCR-T and TIL.
- DMF available
- Buffering ability is strengthened to keep pH stable during culture
The serum-free medium is an updated version and is ideal for serum reduced or serum-free culture conditions. This medium is compatible with following: CIK cells, NK cells, CAR-NK lines, T-cells, CAR-T, TCR-T and TIL.
Both formulations are prepared using select high quality reagents and raw materials including GMP-grade material. Both media are manufactured in certified ISO 9001 and ISO 13485 facilities.
Delivery: Shipped on ice packs.
Specifications
- Storage temperature:4 °C